Learn More
Gilson™ EXPERTTM Universal Fit Pipette Tips
Featuring patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding when dispensing
Supplier:  Gilson™ F1733181
 View more versions of this product
Catalog No. F1733181G
Only null left
Description
Description
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Tips feature graduations at varying volumes allowing for quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Specifications
1 to 200 μL
Non-sterile
Autoclavable
8.255cm
Gel Loading
Rack
Racked
Air Displacement Tips
Single-use
Yes
Polymer
5μL
Non-filtered
Clear
Gel Loading
RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free
0.37 mm
Flat
200/Cs.
Yellow
Universal Fit
EXPERT
Flat
Product Suggestions
Product Suggestions