Learn More
Gilson™ Tip Empty Rack For Pipet Tips
Use with loose Gilson Diamond tips. GilsonTM Tip Empty Rack For Pipette Tips are color coded for ease of use.
Supplier:  Gilson™ F171302
 View more versions of this product
Catalog No. F171302G
Only null left
Description
Description
  • Fully autoclavable
  • Made of polypropylene
Specifications
Specifications
Autoclavable
D200 Pipette Tips (2 to 200μL)
1
Yes, Interlocking
Diamond Tips
134 x 89 x 127 mm
3.66 in.
4.92 in.
Empty Rack
Yellow
Polypropylene
Rectangular
Non-sterile
2
9.3 cm
5.28 in.
2 Empty Racks
1x2
Product Suggestions
Product Suggestions