missing translation for 'onlineSavingsMsg'
Learn More

Giemsa Stain Solution, Certified, LabChem™

$68.73 - $143.12

Specifications

Chemical Name or Material Giemsa Stain Solution
Physical Form Liquid
CAS 56-81-5,67-56-1,51811-82-6
Density 1.03g/mL
CAS Max % 60.98,38.28,0.74
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC148407
View Documents
LabChem
LC148407
125 mL
Each for $68.73
Only null left
 
LC148401
View Documents
LabChem
LC148401
500 mL
Each for $143.12
Only null left
 
Description

Description

Specifications

Specifications

Giemsa Stain Solution
56-81-5,67-56-1,51811-82-6
60.98,38.28,0.74
Soluble in water
Hydrogen bromide, Bromine; Carbon monoxide; Carbon dioxide; Hydrogen chloride; Sulfur compounds
C14H14ClN3S
291.8
Liquid
1.03g/mL
Poly Bottle
1.05g/mL ±0.02
Blue
UN1993
Certified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'