missing translation for 'onlineSavingsMsg'
Learn More

Giemsa Stain Solution, Certified, LabChem™

Supplier:  LabChem LC148401

 View more versions of this product

Catalog No. LC148401

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Liquid
56-81-5,67-56-1,51811-82-6
60.98,38.28,0.74
Soluble in water
Hydrogen bromide, Bromine; Carbon monoxide; Carbon dioxide; Hydrogen chloride; Sulfur compounds
500 mL
UN1993
Certified
Giemsa Stain Solution
1.03g/mL
Poly Bottle
1.05g/mL ±0.02
Blue
C14H14ClN3S
291.8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions