missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giemsa Stain, Certified, Spectrum™ Chemical
$272.64 - $11146.32
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18610710
|
Spectrum Chemical Mfg Cor
GI1201GM |
1 g | Amber Glass Bottle |
Each for $272.64
|
|
||||
|
18610714
|
Spectrum Chemical Mfg Cor
GI1205GM |
5 g | Amber Glass Bottle |
Each for $370.32
|
|
||||
|
18610713
|
Spectrum Chemical Mfg Cor
GI12025GM |
25 g | Amber Glass Bottle |
Each for $1,553.25
|
|
||||
|
18610712
|
Spectrum Chemical Mfg Cor
GI120100GM |
100 g | Amber Glass Bottle |
Each for $1,807.68
|
|
||||
|
18610711
|
Spectrum Chemical Mfg Cor
GI1201KGBL |
1 kg | Amber Glass Bottle |
Each for $4,916.54
|
|
||||
|
18610715
|
Spectrum Chemical Mfg Cor
GI1205KGBL |
5 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 51811-82-6 |
Specifications
| 51811-82-6 | |
| Certified | |
| Blue/Green | |
| Giemsa Stain |
| 1 | |
| Amber Glass Bottle | |
| 1 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.