missing translation for 'onlineSavingsMsg'
Learn More

Giemsa Stain, Certified, Spectrum™ Chemical

Catalog No. 18610715
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610715 5 kg Amber Glass Bottle
18610710 1 g Amber Glass Bottle
18610714 5 g Amber Glass Bottle
18610713 25 g Amber Glass Bottle
18610712 100 g Amber Glass Bottle
18610711 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610715 Supplier Spectrum Chemical Mfg Cor Supplier No. GI1205KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51811-82-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Blue/Green
Quantity 5 kg
Grade Certified
Chemical Name or Material Giemsa Stain

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.