missing translation for 'onlineSavingsMsg'
Learn More

Giemsa's azur-eosin-methylene blue, For Microscopy, MilliporeSigma™

$284.12 - $862.48

Specifications

Chemical Name or Material Giemsa's azur-eosin-methylene blue
CAS 17372-87-1 , 61-73-4
Odor Weak characteristic odor
Packaging Glass bottle
Color Dark Green
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M1092030025
View Documents Promotion Details
N/A 25 g
Each for $284.12
Only null left
 
M1092030100
View Documents Promotion Details
N/A 100 g
Each for $862.48
Only null left
 
Description

Description

For microscopy
Specifications

Specifications

Giemsa's azur-eosin-methylene blue
Weak characteristic odor
Dark Green
17372-87-1 , 61-73-4
Glass bottle
Solid
Videos
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

SDS
Documents

Documents