missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin-EDTA (0.05%), phenol red
GREENER_CHOICE

Catalog No. 25300120
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
20 x 100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25300120 20 x 100 mL
25300062 500 mL
25300054 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25300120 Supplier Gibco™ Supplier No. 25300120

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas.

Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements.

  • Documented traceability
  • Dual-site cGMP Manufacturing
  • Quality testing
  • Solutions tested for pH, osmolality, sterility, and performance
  • Raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
  • Documented traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, viral inactivation summary, and supply chain transparency

Specifications

Chelators EDTA
Content And Storage Storage conditions: -5°C to -20°C
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin
Concentration 1X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red, EDTA
Osmolality 270 - 310 mOsm/kg
pH 7.2 to 8
Product Line Gibco
Quantity 20 x 100 mL
Shipping Condition Wet Ice
Source Animal Origin
Tested For In vitro Bioassay
Show More Show Less
How do you isolate cells from tissue?

Dissociation of cells from Primary Tissue

TRYPSIN
1. After dissecting off unusable tissue, mince the remaining tissue into 3 to 4 mm pieces with a sterile scalpel or scissors. Wash the tissue pieces by resuspending in a balanced salt solution without calcium and magnesium. Allow the tissue pieces to settle, and remove the supernatant. Repeat the wash 2 or 3 times.
2. Place the container with the tissue pieces on ice, and remove any remaining supernatant. Add 0.25% trypsin in a balanced salt solution without calcium or magnesium (1 ml of trypsin for every 100 mg of tissue).
3. Incubate at 4°C for 6 to 18 h to maximize penetration of the enzyme with little trypsin activity.
4. Decant and discard the trypsin from the tissue pieces. Incubate the tissue pieces with residual trypsin at 37°C for 20 to 30 min.
5. Add warm, complete media to the tissue pieces and gently disperse the tissue by pipetting. If using a serum-free medium, also add soybean trypsin inhibitor.
6. Filter the cell suspension through sterile, stainless steel mesh (100 to 200 µm) to completely disperse any remaining tissue. Count and seed the cells for culture.

COLLAGENASE
1. Mince tissue into 3 to 4 mm pieces with a sterile scalpel or scissors. Wash the tissue pieces several times with Hanks' Balanced Salt Solution (HBSS).
2. Add collagenase (50 to 200 U/ml in HBSS).
3. Incubate at 37°C for 4 to 18 h. Addition of 3 mM CaCl2 increases the efficiency of dissociation.
4. Filter the cell suspension through a sterile stainless steel or nylon mesh to separate the dispersed cells and tissue fragments from the larger pieces. Fresh collagenase can be added to the fragments if further disaggregation is required.
5. Wash suspension several times by centrifugation in HBSS.
6. Resuspend the pellet in culture medium. Count and seed the cells for culture.

DISPASE
1. Mince tissue into 3 to 4 mm pieces with a sterile scalpel or scissors. Wash the tissue pieces several times in a calcium and magnesium-free balanced salt solution.
2. Add dispase (0.6 to 2.4 U/ml in calcium and magnesium-free balanced salt solution).
3. Incubate at 37°C for 20 min to several hours.
4. Filter the cell suspension through a sterile, stainless steel or nylon mesh to separate the dispersed cells and tissue fragments from the larger pieces. Fresh dispase can be added to the fragments if further disaggregation is required.
5. Wash suspension several times by centrifugation in the balanced salt solution.
6. Resuspend the pellet in culture medium. Count and seed the cells for culture.

REFERENCE:
Freshney, R. (1987) Culture of Animal Cells: A Manual of Basic Technique, p. 117, Alan R. Liss, Inc., New York.

Does Diploid Growth Serum-Reduced Medium (SRM) contain L-glutamine?

The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.

Do I have to adjust my infection parameters when using Diploid Production Serum-Free Medium (SFM)?

We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.

Can Diploid Growth Serum-Reduced Medium (SRM) be supplemented with different types of serum?

Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.

Which cell lines can I culture in Diploid Growth Serum-Reduced Medium (SRM)?

Diploid Growth Serum-Reduced Medium has been evaluated with fibroblast cell lines, including MRC-5, WI-38, IMR-90, BS-2, and CEF cells.

How long can I use Diploid Growth Serum-Reduced Medium (SRM) supplemented with Diploid Growth Supplement?

Once supplemented, the complete Diploid Growth Serum-Reduced Medium must be used within 4 weeks.

Can I freeze or thaw my cells in Diploid Growth Serum-Reduced Medium (SRM)?

Most diploid cell lines can be frozen and thawed in Diploid Growth Serum-Reduced Medium. Consult the Diploid Growth Serum Reduced Medium User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0019015_DiploidGrowthSerumReducedMedium_UG.pdf) for additional information.

How do I adapt my cells to Diploid Growth Serum-Reduced Medium (SRM)?

Cells growing in medium with serum can be transferred directly to Diploid Growth Serum-Reduced Medium without adaptation. Continue to monitor and passage cells for 2-3 passages until consistent growth is achieved. Cultures may benefit from supplementation with 1-2% serum.

How long can I keep trypsin in the refrigerator after thawing?

Generally speaking, trypsin can be stored refrigerated for up to a week. However, this may vary depending on how often it is warmed up and used during that week. It may also depend on how tightly the cells are attached, because there will be some loss of activity each day the trypsin is not frozen.

What is the difference between TrypLE Express Enzyme, TrypLE Select Enzyme, and CTS TrypLE Select Enzyme?

All the above products contain rProtease, a non-animal, trypsin-like enzyme formulation used for the dissociation of attachment-dependent cell lines. This alternative for porcine trypsin is a recombinant enzyme derived from microbial fermentation.

The TrpLE Express, TrypLE Select, and CTS TrypLE Select products differ in their production and testing methods and in the documentation provided.

TrypLE Select is for the industrial market and has release-testing both for endotoxins and enzyme activity. This material also has a Drug Master File on record. It is manufactured in AOF-dedicated equipment and is intended for Research Use/Further Cell Culture Manufacturing.

TrypLE Express is manufactured without use of dedicated equipment and is intended for Research Use Only. Not for use in diagnostic procedures.

CTS TrypLE Select is manufactured under scalable cGMP conditions using dedicated equipment and is designed for clinical applications. This product has a Drug Master File on record and is intended for Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. Not intended for direct administration into humans or animals.

To read more about TrypLE and cell release times using this reagent, please go here (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin/tryple-express.html#1).

What are the benefits of Gibco TrypLE reagent over trypsin?

TrypLE reagent is free of animal- and human-derived components, producing an exceptionally pure reagent that is gentle on cells. Inactivation with trypsin inhibitors is not required. TrypLE reagent is ideal for dissociating a variety of attachment-dependent mammalian cell lines both in serum and in serum-free conditions, and can be directly substituted for trypsin in your current protocol. TrypLE reagent is room-temperature stable and ready to use when you need it. TrypLE cell dissociation reagents remain stable for 24 months at room temperature, making storage and handling easier and more convenient.

What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).