Learn More
Gibco™ RPMI 1640 Medium, powder, HEPES
Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer.
$134.00 - $191.00
Specifications
| Cell Line | HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas |
|---|---|
| Cell Type | Leukemic Cells |
| Classification | Animal Origin-free |
| Form | Powder |
| Product Type | RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) |
Requires: Supplementation
Description
Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes and carcinomas. Life Technologies offers a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This RPMI is modified as follows:
With: L-glutamine, Phenol Red, HEPES
Without: Sodium Bicarbonate
The complete formulation is available.
Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12 and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference our Type II Drug Master File (DMF).
RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.
Order Info
Shipping Condition: Room Temperature
Specifications
| HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas | |
| Animal Origin-free | |
| RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) | |
| Sterile-filtered | |
| No Sodium Pyruvate, No Sodium Bicarbonate | |
| Gibco | |
| Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
| Leukemic Cells | |
| Powder | |
| Standard Serum Supplementation | |
| Glutamine, HEPES, Phenol Red | |
| cGMP-compliant under the ISO 13485 standard | |
| Room Temperature |
Safety and Handling
ShelfLife : 36 Months From Date of Manufacture
For Research Use Only. Not for use in diagnostic procedures. Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from Life Technologies to reference their Type II Drug Master File (DMF). For in vitro diagnostic use.