missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Chicken Serum, New Zealand Origin
Sustainable packaging
Supplier: Gibco™ 16110082
Description
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
- Sera Category: Specialty
- Origin: New Zealand
- Age: Adult
Cell Culture, Mammalian Cell Culture
Order Info
Shipping condition: Dry ice
Specifications
| Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
| Adult | |
| Liquid | |
| 500 mL | |
| Sterile-filtered | |
| Frozen |
| Chicken | |
| New Zealand | |
| Chicken Serum | |
| Sterile-filtered | |
| Gibco |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.