missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM α, nucleosides, GlutaMAX™ Supplement DFS Item

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells.

$124.00 - $1578.00

Specifications

Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type MEM α (Minimum Essential Medium α)
Serum Level Standard Serum Supplementation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
32571036 DFS Item
View Documents
Gibco™
32571036
500 mL
Each for $124.00
Only null left
 
32571101 DFS Item
View Documents
Gibco™
32571101
10 x 500 mL
Each for $1,578.00
Only null left
 
Description

Description

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM α is modified as follows:

With: Ribonucleosides, Deoxyribonucleosides, GlutaMAX™, Phenol Red

The complete formulation is available.

Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. Gibco™ MEM α with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. This product is made with Earle's salts.

MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
1 X
MEM α (Minimum Essential Medium α)
Sterile-filtered
No HEPES
Gibco, GlutaMAX
Videos
SDS
Documents

Documents

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.