missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM, powder

Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells.

$178.00 - $263.00

Specifications

Cell Line Jurkat, COS-7, and macrophage cells
Classification Animal Origin-free
Form Powder
Product Type IMDM (Iscove's Modified Dulbecco's Medium)
Sterility Sterile-filtered
View More Specs

Requires: Supplementation


Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12200036
Encompass
View Documents Promotion Details
Gibco™
12200036
10 x 1 L
Each for $263.00
Only null left
 
12200069
Encompass
View Documents Promotion Details
Gibco™
12200069
10 L
Each for $178.00
Only null left
 
Description

Description

Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:

With: L-glutamine, Phenol Red

Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate.

Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Jurkat, COS-7, and macrophage cells
Powder
Sterile-filtered
No α-Thioglycerol, No 2-Mercaptoethanol
Gibco
Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Animal Origin-free
IMDM (Iscove's Modified Dulbecco's Medium)
High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
Room Temperature
Videos
Safety and Handling

Safety and Handling

ShelfLife : 36 Months From Date of Manufacture

SDS
Documents

Documents

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.