missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Horse Serum, heat inactivated, New Zealand origin

Catalog No. 26050070
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26050070 100 mL
26050088 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26050070 Supplier Gibco™ Supplier No. 26050070

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EIA tested (negative) serum from a donor herd

  • Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays
  • It has earned the trust of researchers with consistent quality and award-winning support
  • Helps meet research needs and budget requirements
  • EIA tested (negative) serum from a donor herd
  • Shipped frozen, store at <10°C
  • Sera Category: Specialty
  • Origin: New Zealand

Order Info

Shipping Condition: Dry ice

Specifications

Content And Storage Storage conditions: ≤-10°C, Shipping conditions: Frozen
Species Horse/Equine, Horse
Country of Origin New Zealand
Form Liquid
Product Type Horse Serum
Quantity 100 mL
Serum Treatment Heat-inactivated
Sterility Sterile
Product Line Gibco
Shipping Condition Frozen
Does horse serum contain tetracycline?

No. Our horse serum is collected from a controlled herd. Should one of the animals require treatment with an antibiotic, that animal is removed from the drawing program for the full witholding period of any drug administered.

When should I use heat-inactivated serum?

Heat-inactivation of serum inactivates the “complements” in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.