missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gurr Buffer Tablets

Catalog No. 10582013
Encompass
Change view
Click to view available options
Quantity:
50 TabLets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10582013 50 TabLets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10582013 Supplier Gibco™ Supplier No. 10582013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A convenient way to prepare a pH 6.8 phosphate buffer for dilution of Giemsa stain

  • Used in G-banding of chromosomes for cytogenetic analysis
  • Add one buffer tablet to 100 mL of distilled water and stir until dissolved
  • Package size: 50 tablets

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Storage conditions: 15–30°C
Shipping conditions: Ambient

Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Form Powder
Product Type Gurr Buffer
For Use With (Application) Clinical Research, Translational Research
pH 6.8
Quantity 50 TabLets
Shipping Condition Room Temperature
How long can I store Gurr Buffer Tablets (Cat. No. 10582013) that have been reconstituted?

After reconstituting Gurr Buffer Tablets, the subsequent solution can be stored at 4 degrees C for six months.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.