missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Grace's Insect Medium, supplemented
GREENER_CHOICE

Widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21

$142.00 - $1,290.00

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Form Liquid
Product Type Grace's Insect Medium
With Additives Glutamine, Yeastolate, Lactalbumin Hydrolysate
View More Specs

Requires: Grace's Unsupplemented Insect Medium requires ambient CO2 in order to maintain physiological pH.


Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11605094
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
11605094
500 mL
Each for $142.00
Only null left
 
11605102
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
11605102
10 x 500 mL
Each for $1,290.00
Only null left
 
Description

Description

Originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS).

  • Contains lactalbumin hydrolysate, yeastolate, L-glutamine, and sodium bicarbonate
  • Dual site cGMP manufacturing and quality system
  • Commonly supplemented with 10% Fetal Bovine Serum (FBS) before use

Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Sf21, Sf9
Liquid
Glutamine, Yeastolate, Lactalbumin Hydrolysate
Gibco
Calcium, Magnesium
Insect Cell
Grace's Insect Medium
No Insect Hemolymph
Room Temperature
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.