Learn More
Gibco™ FreeStyle™ CHO Expression Medium
Animal origin-free, chemically defined, protein-free medium
$257.00 - $1102.00
Specifications
| Product Type | Expression Medium |
|---|---|
| Cell Line | FreeStyle™ CHO-S Cells, CHO |
| Content And Storage | Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
| Classification | Chemically Defined, Protein-Free, Serum-Free |
| Culture Type | Suspension Cell Culture |
Description
FreeStyle CHO Expression Medium is specifically developed to support the growth and transient transfection of CHO-S cells in suspension without adaptation. This medium requires supplementation with 8 mM glutamine before use.
- Formulation without hypoxanthine and thymidine for use with or without dhfr systems
- Ability to support high-density suspension growth and transient transfection of CHO-S cells
- Animal origin-free, protein-free and chemically defined formulation
- Easy use as part of the FreeStyle MAX CHO Expression System for protein production
- Scalability in spinner flasks and bioreactors
Formulation without hypoxanthine and thymidine for use with or without dhfr systems
- Gibco FreeStyle CHO Expression Medium is made without hypoxanthine and thymidine for use in dihydrofolate reductase (dhfr) amplification systems
- Non-dhfr systems require the addition of HT Supplement before use
- Gibco FreeStyle CHO cells in suspension can be cultured directly into Gibco FreeStyle CHO Expression Medium without adaptation
- Cells adapted to other commercially available serum-free media can be subcultured directly into Gibco FreeStyle CHO Expression Medium, usually without any further adaptation
- Cells usually require adaptation from serum-containing formulations
- Cells cultured in Gibco FreeStyle CHO Expression Medium can be transfected using lipid-based transfection reagents, such as FreeStyle MAX reagent
- Gibco FreeStyle CHO Expression Medium is animal origin–free, protein-free, and chemically defined, allowing for easier purification of your protein of interest
- Gibco chemically defined media contain no proteins, hydrolysates, or components of unknown composition
- Protein production using Gibco FreeStyle CHO Expression Medium can be scaled up in spinner flasks or bioreactors
- The appropriate spinner or impeller speed and seeding density should be optimized for each system
- Depending on the impeller design and speed, it may be necessary to supplement Gibco FreeStyle CHO Expression Medium with additional Pluronic™ F-68 to avoid sheer stress in the culture
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room temperature
Compliance
Gibco FreeStyle CHO Expression Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
| Expression Medium | |
| Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
| Suspension Cell Culture | |
| No Glutamine | |
| Serum-Free | |
| Liquid |
| FreeStyle™ CHO-S Cells, CHO | |
| Chemically Defined, Protein-Free, Serum-Free | |
| ≤0.25 EU/mL, ≤ 0.25 EU/mL | |
| No Phenol Red | |
| Freestyle, Gibco |
For Research Use Only. Not for use in diagnostic procedures.