missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, New Zealand

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

$2,850.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Country of Origin New Zealand
Form Liquid
Product Type Fetal Bovine Serum
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
10091155
Encompass
View Documents Promotion Details
Gibco™
10091155
1000 mL Bottle
Each for $2,850.00
Only null left
 
Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Premium (Secure)
  • Origin: New Zealand
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤30 mg/dL (levels routinely ≤25 mg/dL)

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
New Zealand
Fetal Bovine Serum
Sterile
≤30 mg/dL
Fetal
Liquid
Standard (Sterile-filtered)
≤5 EU/mL
Frozen
Videos
SDS
Documents

Documents

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.