Learn More
Gibco™ Embryonic stem-cell FBS, qualified, USDA-approved regions
Embryonic stem-cell FBS, qualified, USDA-approved regions is US certified performance plus sera for broad range of cell types, especially sensitive cell lines.
$1,454.00
Specifications
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
|---|---|
| Species | Cattle/Bovine |
| Age | Fetal |
| Country of Origin | Mexico |
| Form | Liquid |
Description
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Origin: Countries meeting USDA importation requirements
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells.
Order Info
Shipping Condition: Dry Ice
Specifications
| Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
| Fetal | |
| Liquid | |
| Standard (Sterile-filtered) | |
| <50 EU/mL | |
| ES Cell-Qualified |
| Cattle/Bovine | |
| Mexico | |
| Fetal Bovine Serum | |
| Sterile | |
| ≤25 mg/dL | |
| Frozen |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.