missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Trypsin Neutralizer Solution
GREENER_CHOICE

Catalog No. R002100
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R002100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R002100 Supplier Gibco™ Supplier No. R002100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sterile, phosphate-buffered saline solution

Contains (1X) containing calf serum as a trypsin inhibitor.

Sustainable packaging

Cell Culture, Corneal Epithelial Cell Culture, Fibroblast Culture, Keratinocyte Culture, Large Vessel Endothelial Cell Culture, Mammalian Cell Culture, Mammary Epithelial Cell Culture, Melanocyte Culture, Microvascular Endothelial Cell Culture, Primary Cell Culture, Smooth Muscle Cell Culture

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage minus 20 degrees C
Cell Type Mammalian Cells
Concentration 1 X
Product Type Trypsin inhibitor
Sterility Sterile-filtered
pH 7.4
Product Line Cascade Biologics
Quantity 100 mL
Shipping Condition Room Temperature
What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

For Research Use Only. Not for use in diagnostic procedures.