missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gibco™ Bottle, 1000 mL
GREENER_CHOICE

Catalog No. 10341001
Encompass
Change view
Click to view available options
Quantity:
60 bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10341001 60 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10341001 Supplier Invitrogen™ Supplier No. 10341001

Gibco™ Bottle, 1000 mL

Sterile, empty GIBCO 1000 mL bottles for use in cell culture research

Specifications

Capacity (Metric) 1 L
Material PET
Quantity 60 bottles
Sterility Sterile
Product Line Gibco
Content And Storage Sterile 60 x 1000 mL bottles irradiated to 10^-6 sterility. Store at RT
Shipping Condition Room Temperature
Mouth Wide
Product Type Media Bottle

For Research Use Only. Not for use in diagnostic procedures.