missing translation for 'onlineSavingsMsg'
Learn More

Gibberellin A3 98.0+%, TCI America™

Catalog No. p-7130107
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0029100MG 100 mg
G00291G 1 g
G00295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0029100MG Supplier TCI America Supplier No. G0029100MG

Chemical Identifiers

CAS 77-06-5
Molecular Formula C19H22O6
Molecular Weight (g/mol) 346.38
MDL Number MFCD00079329
InChI Key IXORZMNAPKEEDV-QTWFBFKQSA-N
Synonym gibberellic acid, gibberellin a3, gibberellin, gibreskol, brellin, cekugib, grocel, gibberellin x, gibberellic acid ga3, gib-tabs
PubChem CID 91757643
IUPAC Name (1R,2R,5S,8S,9S,10R,12S)-5,12-dihydroxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.1⁵,⁸.0¹,¹⁰.0²,⁸]heptadec-13-ene-9-carboxylic acid
SMILES CC12[C@H]3[C@H](C(O)=O)[C@@]45CC(=C)[C@@](O)(C4)CC[C@H]5[C@]3(OC1=O)C=C[C@@H]2O

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 100 mg
Formula Weight 346.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Gibberellin A3
EINECSNumber (4)-0785&(9)-2556
RTECSNumber LY8990000
TSCA Yes