missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, Crystal, EP, BP, JP, USP, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18600906
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600906 50 kg Poly Pail
18600902 500 g Poly Pail
18600903 2.5 kg Poly Pail
18600904 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600906 Supplier Spectrum Chemical Mfg Cor Supplier No. BE14050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BE140, 65-85-0, C7H6O2

Spectrum™ Chemical Benzoic Acid, Crystal, USP, EP, BP, JP has antifungal and topical antiseptic properties. Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
IUPAC Name benzoic acid
SMILES OC(=O)C1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Loss on Drying 0.5%
Packaging Poly Pail
Quantity 50 kg
Percent Purity 99.5 to 100.5%
Grade Multi Compendia
Physical Form Crystalline Powder or Lumps