missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFPT2 Recombinant Rabbit Monoclonal Antibody (K01_1R65)
GREENER_CHOICE

Catalog No. MA563644
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA563644 100 μL
MA565103 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA563644 Supplier Invitrogen™ Supplier No. MA563644
Only null left

Rabbit Recombinant Monoclonal Antibody

GFPT2 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

GFPT2 controls the flux of glucose into the hexosamine pathway. This protein most likely involved in regulating the availability of precursors for N-and O-linked glycosylation of proteins.

Specifications

Antigen GFPT2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1R65
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene GFPT2
Gene Accession No. O94808, Q4KMC4, Q9Z2Z9
Gene Alias AI480523; D-fructose-6-phosphate amidotransferase 2; GFAT; GFAT 2; GFAT2; GFPT2; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine fructose-6-phosphate transaminase 2; glutamine: fructose-6-phosphate aminotransferase 2; glutamine:fructose 6 phosphate amidotransferase 2; glutamine:fructose-6-phosphate amidotransferase 2; glutamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine-fructose-6-phosphate transaminase 2; Hexosephosphate aminotransferase 2
Gene Symbols GFPT2
Host Species Rabbit
Immunogen Recombinant protein of human GFPT2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14584, 360518, 9945
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less