missing translation for 'onlineSavingsMsg'
Learn More

GFP APC-conjugated Antibody, R&D Systems™

Catalog No. IC4240A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4240A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC4240A Supplier Bio-Techne Supplier No. IC4240A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Goat Polyclonal Antibody

GFP Polyclonal specifically detects GFP in Human∣Multi-species samples. It is validated for Flow Cytometry∣Flow Cytometry (Intracellular Staining).

Specifications

Antigen GFP
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P42212
Gene Alias eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp)
Gene Symbols GFP
Host Species Goat
Immunogen E. coli-derived recombinant GFPuv Ser2-Lys238 Accession # P42212
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects GFP in direct ELISAs and Western blots.
Target Species Human, Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less