missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002670B01P
Description
Sequence: MERRRITSAARRSYVSSGEMMVGGLAPGRRLGPGTRLSLARMPPPLPTRVDFSLAGALNAGFKETRASERAEMMELNDRFASYIEKVRFLEQQNKALAAELNQLRAKEPTKLADVYQAELRELRLRLDQLTANSARLEVERDNLAQDLATVRQKLQDETNLRLEAENNLAAYRQEADEATLARLDLERKIESLEEEIRFLRKIHEEEVRELQEQLARQQVHVELDVAKPDLTAALKEIRTQYEAMASSNMHEAEEWYRSKFADLTDAAARNAELLRQAKHEANDYRRQLQSLTCDLESLRGTNESLERQMREQEERHVREAASYQEALARLEEEGQSLKDEMARHLQEYQDLLNVKLALDIEIATYRKLLEGEENRITIPVQTFSNLQIRETSLDTKSVSEGHLKRNIVVKTVEMRDGEVIKESKQEHKDVMSpecifications
| GFAP | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_002055.2 | |
| GFAP | |
| GFAP (NP_002046.1, 1 a.a. ∼ 432 a.a) full-length human protein. | |
| RUO | |
| 2670 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| glial fibrillary acidic protein | |
| FLJ45472 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |