Learn More
GFAP (Glial Fibrillary Acidic Protein) Rabbit Polyclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ GFAP (Glial Fibrillary Acidic Protein), Rabbit Polyclonal Antibody.
Supplier: Thermo Fisher Scientific RB087A
Description
Glial Fibrillary Acidic Protein (GFAP) is specific to astrocytes (i.e., glial cells) and ependymal cells of the central nervous system.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Cow, Sheep, Guinea Pig, Chicken, Mouse, Rat and Hamster. Others not known.
Epitope: Not determined
Immunogen: GFAP isolated from cow spinal cord
Molecular Weight: 51-52 kDa
Positive Control: Brain or astrocytoma
Cellular Localization: Cytoplasmic Ab-4 stains astrocytes, glial cells, ependymal cells and their corresponding tumors. In the peripheral nervous system, Schwann cells, satellite cells, and enteric glial cells are stained. Weak staining of axons has been observed. Absorption studies show that this is caused by a cross-reaction with neurofilament protein. Ab-4 is particularly useful for distinguishing neoplasms of astrocytic origin from other neoplasms in the central nervous system by immunohistochemistry.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| GFAP (Glial Fibrillary Acidic Protein) | |
| Polyclonal | |
| Rabbit | |
| Brain or astrocytoma | |
| RUO | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| GFAP isolated from cow spinal cord | |
| 1 mL | |
| Neuroscience | |
| Bovine, Chicken, Guinea Pig, Hamster, Human, Mouse, Non-human Primate, Rat | |
| IgG |