missing translation for 'onlineSavingsMsg'
Learn More

Gestodene, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 468650010

 View more versions of this product

Catalog No. AC468650010


Only null left

Description

Description

Specifications

Specifications

Gestodene
Available
Powder
00867858
197.9°C
13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one
CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C
310.44
310.43
Glass Bottle
60282-87-3
C21H26O2
White to Beige
4425
SIGSPDASOTUPFS-KQMXEUTGSA-N
(1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
1 g

Chemical Identifiers

60282-87-3
310.44
SIGSPDASOTUPFS-KQMXEUTGSA-N
(1R)-11a-ethyl-1-ethynyl-1-hydroxy-1H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
C21H26O2
00867858
13beta-Ethyl-17beta-hydroxy-18,19-dinorpregna-4,15-dien-20-yn-3-one
CCC12CCC3C(CCC4=CC(=O)CCC34)C1C=C[C@@]2(O)C#C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS08: Health hazard

ShelfLife : 5 years

EINECSNumber : 262-145-8

RTECSNumber : JF7956000

Recommended Storage : Freezer -20°C

SDS
Documents

Documents

Promotions

Promotions