missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germanium(IV) oxide, 99.99% (metals basis)
Description
Used in phosphors, transistors and diodes, and infrared transmitting glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1086°C |
| Color | White |
| Physical Form | Sintered Lump |
| Quantity | 2 g |
| Assay Percent Range | 99.99% (Metals basis) |
| Solubility Information | soluble in water; insoluble in HF and HCl. |
| Formula Weight | 104.59 |
| Odor | Odorless |
| Density | 6.239 g/cm3 |
| Chemical Name or Material | Germanium(IV) oxide |
Safety and Handling
| Hazard Category | H302+H332 |
| Hazard Statement | Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| EINECSNumber | 215-180-8 |
| RTECSNumber | LY5240000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only