missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) oxide, 99.99% (metals basis)

Catalog No. AA3638204
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3638204 2 g
AA3638209 10 g
AA3638218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3638204 Supplier Thermo Scientific Chemicals Supplier No. 03638204

Used in Phosphors, Transistors and Diodes, and Infrared Transmitting Glass | CAS: 1310-53-8 | GeO2 | 104.628 g/mol

Used in phosphors, transistors and diodes, and infrared transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
MDL Number MFCD00011030
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
Synonym germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O

Specifications

Melting Point 1086°C
Color White
Physical Form Sintered Lump
Quantity 2 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information soluble in water; insoluble in HF and HCl.
Formula Weight 104.59
Odor Odorless
Density 6.239 g/cm3
Chemical Name or Material Germanium(IV) oxide
Hazard Category H302+H332
Hazard Statement Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 215-180-8
RTECSNumber LY5240000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only