missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germanium(IV) ethoxide, 99.995% (metals basis)
Description
Preparation of Germanium oxide by CVD.Sol-gel intermediate for optical fibers and photosensitive SiO2-BeO2 glasses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -72°C |
| Color | Colorless |
| Density | 1.14 g/mL |
| Boiling Point | 185.5°C |
| Flash Point | 49°C (121°F) |
| Odor | Alcohol-like |
| Refractive Index | 1.407 |
| Linear Formula | Ge(OC2H5)4 |
| Quantity | 5 g |
| UN Number | UN1993 |
| Show More |
Safety and Handling
| Hazard Category | H226-H301+H311+H331-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H315-H319 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 238-007-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only