missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) ethoxide, 97%

Catalog No. p-7049553
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1467903 1 g
AA1467906 5 g
AA1467914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1467903 Supplier Thermo Scientific Chemicals Supplier No. 01467903
Only null left

CAS: 14165-55-0

It is a sol-gel intermediate for optical fibers and photosensitive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14165-55-0
MDL Number MFCD00015121
Synonym Germanium tetraethoxide; Tetraethoxygermane

Specifications

Melting Point -72°C
Color Colorless
Density 1.14 g/mL
Boiling Point 185.5°C
Flash Point 49°C (121°F)
Odor Alcohol-like
Refractive Index 1.407
Linear Formula Ge(OC2H5)4
Quantity 1 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Soluble in anhydrous alcohol and benzene; Hydrolyzes in water
Formula Weight 252.84
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Germanium(IV) ethoxide
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 238-007-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only