missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germanium ICP Standard, 1000 ppm Ge in H2O/tr HF, Ricca Chemical
Supplier: Ricca Chemical Company PGE1KW100
Specifications
| Germanium ICP Standard | |
| 16962-47-3,7664-39-3,7664-39-3 | |
| 1,000ppm Ge | |
| 1 mL = 1 mg Ge | |
| Colorless | |
| 100°C | |
| Miscible with water | |
| Standard | |
| Laboratory |
| Natural LDPE Bottle | |
| 98.0,0.3,0.05 | |
| 100.0,0.31,0.05 | |
| Liquid | |
| 0.0°C | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| 100 mL |
Chemical Identifiers
| 16962-47-3 |