missing translation for 'onlineSavingsMsg'
Learn More

Germacrone 98.0+%, TCI America™

Catalog No. p-7130280
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G051920MG 20 mg
G0519100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G051920MG Supplier TCI America Supplier No. G051920MG

Chemical Identifiers

CAS 6902-91-6
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00210050
InChI Key CAULGCQHVOVVRN-SWZPTJTJSA-N
Synonym (E,E)-Germacra-3,7(11),9-trien-6-one, trans,trans-3,7-Dimethyl-10-isopropylidene-3,7-cyclodecadien-1-one, (3E,7E)-3,7-Dimethyl-10-(propan-2-ylidene)cyclodeca-3,7-dienone
PubChem CID 6436348
IUPAC Name (3E,7E)-3,7-dimethyl-10-(propan-2-ylidene)cyclodeca-3,7-dien-1-one
SMILES CC(C)=C1C\C=C(C)\CC\C=C(C)\CC1=O

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 20 mg
Formula Weight 218.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Germacrone
RTECSNumber GU2210000
TSCA No
Recommended Storage Refrigerator