accessibility menu, dialog, popup

UserName

CAS RN 6902-91-6

IUPAC Name: (3E,7E)-3,7-dimethyl-10-(propan-2-ylidene)cyclodeca-3,7-dien-1-one
Synonyms: germacrone
Molecular Weight (g/mol): 218.34
Molecular Formula: C15H22O
InChi Key: CAULGCQHVOVVRN-SWZPTJTJSA-N
SMILES: CC(C)=C1C\C=C(C)\CC\C=C(C)\CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results