missing translation for 'onlineSavingsMsg'
Learn More

Germacrone 98.0+%, TCI America™

Catalog No. G0519100MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0519100MG 100 mg
G051920MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0519100MG Supplier TCI America Supplier No. G0519100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6902-91-6
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00210050
InChI Key CAULGCQHVOVVRN-SWZPTJTJSA-N
Synonym (E,E)-Germacra-3,7(11),9-trien-6-one, trans,trans-3,7-Dimethyl-10-isopropylidene-3,7-cyclodecadien-1-one, (3E,7E)-3,7-Dimethyl-10-(propan-2-ylidene)cyclodeca-3,7-dienone
PubChem CID 6436348
IUPAC Name (3E,7E)-3,7-dimethyl-10-(propan-2-ylidene)cyclodeca-3,7-dien-1-one
SMILES CC(C)=C1C\C=C(C)\CC\C=C(C)\CC1=O

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 100 mg
Formula Weight 218.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Germacrone
RTECSNumber GU2210000
TSCA No
Recommended Storage Refrigerator