missing translation for 'onlineSavingsMsg'
Learn More

Geranyl Tiglate 98.0+%, TCI America™

Catalog No. G04305ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04305ML 5 mL
G043025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04305ML Supplier TCI America Supplier No. G04305ML
Only null left

Chemical Identifiers

CAS 7785-33-3
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.36
MDL Number MFCD00016622
InChI Key OGHBUHJLMHQMHS-KRDNBFTESA-N
Synonym Tiglic Acid Geranyl Ester
PubChem CID 5367785
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-yl (2E)-2-methylbut-2-enoate
SMILES C\C=C(/C)C(=O)OC\C=C(/C)CCC=C(C)C

Specifications

Color Yellow
Boiling Point 151°C
Quantity 5 mL
Formula Weight 236.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Geranyl Tiglate
EINECSNumber (2)-0970
RTECSNumber RG5927900
TSCA Yes