accessibility menu, dialog, popup

UserName

CAS RN 7785-33-3

CH3CH3OOH3CH3CH3C(E)(E)

CAS RN 7785-33-3

IUPAC Name: (2E)-3,7-dimethylocta-2,6-dien-1-yl (2E)-2-methylbut-2-enoate
Synonyms: geranyl tiglate
Molecular Weight (g/mol): 236.36
Molecular Formula: C15H24O2
InChi Key: OGHBUHJLMHQMHS-KRDNBFTESA-N
SMILES: C\C=C(/C)C(=O)OC\C=C(/C)CCC=C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results