missing translation for 'onlineSavingsMsg'
Learn More

Geraniol 96.0+%, TCI America™

Supplier:  TCI America G0027100ML

 View more versions of this product

Catalog No. G0027100ML


Only null left

Description

Description

Specifications

Specifications

Geraniol
-15°C
230°C
MFCD00002917
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
CC(=CCCC(=CCO)C)C
154.253
CHEBI:17447
≥96.0% (GC)
106-24-1
Yellow
C10H18O
100 mL
GLZPCOQZEFWAFX-JXMROGBWSA-N
(2E)-3,7-dimethylocta-2,6-dien-1-ol
637566
154.25
Liquid

Chemical Identifiers

106-24-1
154.253
GLZPCOQZEFWAFX-JXMROGBWSA-N
637566
(2E)-3,7-dimethylocta-2,6-dien-1-ol
C10H18O
MFCD00002917
geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
CHEBI:17447
CC(=CCCC(=CCO)C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0258

RTECSNumber : RG5830000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions