missing translation for 'onlineSavingsMsg'
Learn More

Geraniol 96.0+%, TCI America™

Catalog No. p-7130105
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G002725ML 25 mL
G0027100ML 100 mL
G0027500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G002725ML Supplier TCI America Supplier No. G002725ML

Chemical Identifiers

CAS 106-24-1
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00002917
InChI Key GLZPCOQZEFWAFX-JXMROGBWSA-N
Synonym geraniol, lemonol, geranyl alcohol, trans-geraniol, e-geraniol, e-nerol, trans-3,7-dimethyl-2,6-octadien-1-ol, e-3,7-dimethylocta-2,6-dien-1-ol, 2e-3,7-dimethylocta-2,6-dien-1-ol, geraniol extra
PubChem CID 637566
ChEBI CHEBI:17447
IUPAC Name (2E)-3,7-dimethylocta-2,6-dien-1-ol
SMILES CC(=CCCC(=CCO)C)C

Specifications

Melting Point -15°C
Color Yellow
Boiling Point 230°C
Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Geraniol
EINECSNumber (2)-0258
RTECSNumber RG5830000
TSCA Yes