missing translation for 'onlineSavingsMsg'
Learn More

Geraniol, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012390
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012390 5 mL
111012389 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012390 Supplier Merck Emd Millipore Supplier No. 487985ML
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 106-24-1
Vapor Density 5.31 (vs. air)
Density 0.879 g/mL (at 20°C)
Flash Point 108°C
Physical Form Neat
Linear Formula (CH3)2 C=CHCH2 CH2 C(CH3)=CHCH2 OH
MDL Number MFCD00002917
Boiling Point 229°C to 230°C
Synonym trans-3,7-Dimethyl-2,6-octadien-1-ol
Type Analytical Standard
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less