missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentian Violet, 96%, Spectrum™ Chemical
G1006, 548-62-9, C25H30ClN3
$239.73 - $3020.50
Chemical Identifiers
| CAS | 548-62-9 |
|---|---|
| Molecular Formula | C25H30ClN3 |
| Molecular Weight (g/mol) | 407.99 |
| InChI Key | ZXJXZNDDNMQXFV-UHFFFAOYSA-M |
| IUPAC Name | tris[4-(dimethylamino)phenyl]methylium chloride |
| SMILES | [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602901
|
Spectrum Chemical Mfg Cor
G100625GM |
25 g | Amber Glass Bottle |
Each for $239.73
|
|
||||
|
18602902
|
Spectrum Chemical Mfg Cor
G1006125GM |
125 g | Amber Glass Bottle |
Each for $417.93
|
|
||||
|
18602903
|
Spectrum Chemical Mfg Cor
G10061KGBL |
1 kg | Amber Glass Bottle |
Each for $3,020.50
|
|
||||
Description
Spectrum™ Chemical Gentian Violet , also known as crystal violet, is a triarylmethane dye that is used as a histological stain. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 548-62-9 | |
| 407.99 | |
| tris[4-(dimethylamino)phenyl]methylium chloride |
| C25H30ClN3 | |
| ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
| [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C |
Specifications
| 548-62-9 | |
| C25H30ClN3 | |
| [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C | |
| 407.99 | |
| Reagent | |
| Dark Green | |
| Gentian Violet |
| 100% | |
| ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
| tris[4-(dimethylamino)phenyl]methylium chloride | |
| 96% | |
| Amber Glass Bottle | |
| 25 g |