missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentian Violet, 2% (w/v) in Methanol, Ricca Chemical
Chemical Identifiers
| CAS | 548-62-9 , 67-56-1 |
|---|---|
| Molecular Formula | C25H30ClN3 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | ZXJXZNDDNMQXFV-UHFFFAOYSA-M |
| PubChem CID | 11057 |
| ChEBI | CHEBI:41688 |
| IUPAC Name | [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
| SMILES | CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |
Chemical Identifiers
| 548-62-9 , 67-56-1 | |
| Mixture | |
| 11057 | |
| [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
| C25H30ClN3 | |
| ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
| CHEBI:41688 | |
| CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |
Specifications
| 548-62-9 , 67-56-1 | |
| 99.0, 2.53 | |
| C25H30ClN3 | |
| CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
| Mixture | |
| CHEBI:41688 | |
| Indicator | |
| Violet | |
| Gentian Violet |
| 96.0, 2.43 | |
| 2% (w/v) | |
| ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
| [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
| 11057 | |
| 2% W/V | |
| Methanolic Solution | |
| Liquid |