missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gentamicin sulfate

Catalog No. AC455310250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455310250 25 g
AC455310010 1 g
AC455310050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC455310250 Supplier Thermo Scientific Chemicals Supplier No. 455310250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gentamicin sulfate, CAS # 1405-41-0, is an aminoglycoside mainly used to inhibit the growth of Gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Gentamicin sulfate is a bactericidal aminoglycoside antibiotic which shows activity against Gram-negative bacteria
  • It works by inhibiting protein biosynthesis through binding the bacterial ribosome 30S subunit

Applications

  • Gentamicin sulfate has been used in studies to determine wound healing due to its antibiotic and antifungal properties
  • It has laboratory research applications in the study of a number of clinical conditions, including actinomycotic infection, appendicitis, cystic fibrosis, osteomyelitis, pelvic inflammatory disease, plague, gonorrhea, and tularemia

Chemical Identifiers

CAS 1405-41-0
Synonym Garamycin, Gentamicin Sulfate

Specifications

Type Gentamicin Sulfate
Melting Point 218.0°C to 237.0°C
Infrared Spectrum Authentic
Water 15% max.
Quantity 25 g
Solubility Information Solubility in water: soluble.
Percent Purity ≥590 IU/mg (Gentamycin)
Chemical Name or Material Gentamicin sulfate
Hazard Category Respiratory sensitisation (category 1), Skin sensitisation (category 1)
Hazard Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction

RUO – Research Use Only