missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gentamicin Monoclonal Antibody (GB73.3), Invitrogen™

Catalog No. PIMA546131
Change view
Click to view available options
Quantity:
100 μg
500 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546131 1 mg
PIMA546053 100 μg
01671955 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIMA546131 Supplier Invitrogen™ Supplier No. MA546131
Only null left

Mouse Monoclonal Antibody

MA1-10725 detects Gentamicin. MA1-10725 has been successfully used in ELISA applications. The MA1-10725 immunogen is Gentamicin, conjugated to KLH.

Gentamicin is an aminoglycoside antibiotic, used to treat many types of bacterial infections, particularly those caused by Gram-negative bacteria. However, gentamicin is not used for Neisseria gonorrhoeae, Neisseria meningitidis or Legionella pneumophila bacterial infections (because of the risk of the patient going into shock from lipid A endotoxin found in certain gram negative organisms).

Specifications

Antigen Gentamicin
Applications ELISA
Classification Monoclonal
Clone GB73.3
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Gentacycol; Garamycin; Gentavet; Gentamicinum; C21H43N5O7
Host Species Mouse
Immunogen Gentamicin, conjugated to KLH.
Purification Method Protein G
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Chemical
Show More Show Less