missing translation for 'onlineSavingsMsg'
Learn More

Amsbio LLC (formerly Genlantis) X-Gal Staining Kit

Catalog No. A10300K
Encompass
Change view
Click to view available options
Quantity:
50 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10300K 50 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10300K Supplier Amsbio LLC (formerly Genlantis) Supplier No. A10300K

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapid and simple assay for determining transfection efficiency with a beta-galactosidase expressing plasmid

  • Sensitive detection of -galactosidase activity in situ
  • Complete ready-to-use assay system containing all the necessary reagents
  • Results in less than 24 hours post-transfection
  • Easy visualization of positive cells
  • Direct assay of transfections in cell culture dishes or in tissues in vivo
  • Designed for sensitive and low-background in situ detection of cells transfected with vectors carrying the lacZ gene

Specifications

Product Type X-Gal staining kit
Sufficient For 50 Assays in 60mm dish
For Use With (Application) Determining transfection efficiency with a beta-galacto sidase expressing plasmid
Kit Contents 125 mL fixing buffer, 125 mL staining buffer, 25X X-Gal stock (4 x 1 mL), 75 mL 10X PBS
Quantity 50 Assays
Storage Requirements -20°C