missing translation for 'onlineSavingsMsg'
Learn More

Amsbio LLC (formerly Genlantis) SoluLyse™ Bacterial Protein Extraction Reagent

Catalog No. L100500
Encompass
Change view
Click to view available options
Quantity:
125 mL
500 mL
Formulation:
20mM Tris HCl Buffer, pH 7.5
50mM MNaHPO4, pH 7.4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
L100500 500 mL 50mM MNaHPO4, pH 7.4
L200125 125 mL 20mM Tris HCl Buffer, pH 7.5
L200500 500 mL 20mM Tris HCl Buffer, pH 7.5
L100125 125 mL 50mM MNaHPO4, pH 7.4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. L100500 Supplier Amsbio LLC (formerly Genlantis) Supplier No. L100500
Only null left

Efficient and gentle extraction of soluble proteins from bacteria

  • Produces high yield of soluble proteins
  • Simple one-step protocol
  • Proprietary formulation of nonionic detergents optimized for bacterial protein extraction
  • Proteins retain more activity
  • Compatible with protease inhibitors, salts, chelating agents and reducing agents
  • Choice of Phosphate or Tris buffer formulations to avoid interference with affinity purification resins and other downstream applications

Specifications

Formulation 50mM MNaHPO4, pH 7.4
Quantity 500 mL
Product Type Bacterial Protein Extraction Reagent
For Use With (Application) Proprietary formulation of nonionic detergents optimized for bacterial Protein extraction, high Protein yield with simple one-step proto col
Content And Storage Room Temperature