Learn More
Genlantis™ SMARTCells™ Competent Cells
Competent E. coli for the highest transformation efficiency of ligated DNA
Supplier: Genlantis™ C101020
Description
- Optimized for difficult-to-transform ligated DNA
- Ideal for all miniprep DNA experiments
- Obtain maximum efficiencies at different DNA concentrations
- Easy-to-use format and protocol
- Provides the high transformation efficiencies using less-than-ideal conditions of post-ligation DNA
Specifications
| Highest transformation efficiency of ligated DNA | |
| 20 x 50μL SmartCells™ chemically competent E. coli (>1 x 109cfu/μg), 6mL SOC medium, 20μL (10pg/μL) pUC19 positive control plasmid | |
| 20 x 50 μL |