missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ PrimaPure™ Preadipocytes Kits
High-quality human preadipocytes from adult heart or subcutaneous tissue
$3,417.58
Specifications
| Content And Storage | 4° and -20°C |
|---|---|
| For Use With (Application) | Study insulin-stimulated glucose transport, growth hormone enhanced lypolysis and obese gene expression |
| Includes | One ampoule of cryopreserved HPAd, 500mL of Human preadipocyte growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution) |
| Media Recommendation | Human Preadipocyte Growth Medium |
| Quantity | 1 Kit |
Specifications
| 4° and -20°C | |
| One ampoule of cryopreserved HPAd, 500mL of Human preadipocyte growth medium, one subculture reagent kit (100mL each of HEPES buffered saline solution, trypsin/EDTA, and trypsin neutralizing solution) | |
| 1 Kit | |
| 16 Population Doublings |
| Study insulin-stimulated glucose transport, growth hormone enhanced lypolysis and obese gene expression | |
| Human Preadipocyte Growth Medium | |
| Human Adipose Tissue |