Learn More
Genlantis™ Mouse Neural Stem Cells (MNSC)
Stem cells are multipotent and can be induced to differentiate into neurons, astrocytes and oligodendrocytes
$4002.92 - $5184.71
Specifications
| Content And Storage | 4°C |
|---|---|
| Species | Mouse |
| For Use With (Application) | In-vitro research use only |
| Quantity | >500000 Cells |
Includes: Complete System contains an ampule of cryopreserved cells (> 500,000 cells), 250 mL of Neural Stem Cell Growth Medium, and two 10cm plates for neurosphere culture.
Description
PrimaPure Rat and Mouse Neural Stem Cells (NSC) are isolated from cerebral striatum and mesencephalon of E14 fetal mouse and E18 fetal rat brain. Additionally, stem cells are available from adult rat brain. These cells are maintained in an undifferentiated proliferative state by culturing them as free-floating neurospheres in serum-free medium optimized with growth factors. Neural stem cells are a source of donor tissue for prospective neural cell therapy research given the ability of neurosphere to retain multi-lineage potential and proliferative ability in vitro.
- Cells are cryopreserved at second passage and can be cultured and propagated for one to two passages prior to induction of phenotypic differentiation
- Each lot functionally characterized
- Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
Specifications
| 4°C | |
| Mouse | |
| In-vitro research use only | |
| >500000 Cells |