missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ FACSmax Cell Dissociation Solution

Catalog No. p-3826516
Encompass
Change view
Click to view available options
Quantity:
10 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T200110 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T200110 Supplier Genlantis™ Supplier No. T200110
Only null left

FACSmax solution is a gentle and highly effective cell dissociation solution. This proprietary formulation of proteolytic, collagenolytic and DNAse enzymes is highly effective in creating single cell suspensions from clumped cell cultures for accurate cell counting, flow cytometry, viral transfection assays, cell sorting, and bioreactor scale-up. Available in ready to use 100mL bottles.

  • Dissociates clumped cells in minutes
  • Results in homogeneous single cell suspension
  • Gentle cell disaggregation for maximum cell viability
  • Yields accurate, reproducible cell counts
  • Saves time. No need for extra PBS washing steps
  • Ready to use solution

Specifications

Content And Storage -20°C
Product Type Cell Culture Dissociation Reagent
Quantity 10 x 100 mL